To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 27
- Clonality:Monoclonal
- Clone:1E1H5
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:IL27
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:IL27p28|IL-27A|IL27A|IL-27|IL30|p28
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. no.:10088-824
- Supplier No.:66164-1-IG
Specifications
About this item
Western Blot: Recombinant Protein, 1:2000-1:20000; IHC: Human Placenta Tissue, 1:20-1:200
Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 1E1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human