To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:BMI1 Proto-Oncogene, Polycomb Ring Finger
- Clonality:Monoclonal
- Clone:2E10A11
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Antigen symbol:BMI1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:ring finger protein 51|murine leukemia viral (bmi-1) oncogene homolog|flvi-2/bmi-1|polycomb group RING finger protein 4|polycomb group protein Bmi1|RNF51|B lymphoma Mo-MLV insertion region 1 homolog|FLVI2/BMI1|PCGF4|RP11-573G6.1|polycomb complex protein BMI-1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. no.:10083-286
- Supplier No.:66161-1-IG
Specifications
About this item
BMI- 1 is one of polycomb group genes, which together with Ring1 strongly enhances the E3 ubiquitin ligase activity of the Ring2 catalytic subunit. Bmi1 plays an important role in the regulation of cell proliferation and senescence through repression of the p16Ink4a and p19Arf genes and is required for maintenance of adult hematopoietic and neural stem cells. The antibody reacts with the 37kd BMI1 protein.
Western Blot:U-937 Cells, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200; IF:COS-7 Cells, 1:10-1:100; IP: HEK-293 Cells, 1:500-1:5000
Type: Primary
Antigen: BMI1
Clonality: Monoclonal
Clone: 2E10A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse