To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nicotinate Phosphoribosyltransferase Domain Containing 1
- Clonality:Monoclonal
- Clone:5D8H10
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Antigen symbol:NAPRT
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:NAPRT1|PP3856
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. no.:10090-806
- Supplier No.:66159-1-IG
Specifications
About this item
Nicotinic acid (NA) is a coenzyme in cellular redox reactions, and is an essential component of metabolic pathways in all living cells. NAPRT1 (Nicotinate phosphoribosyltransferase) is essential for increasing cellular NAD levels and, thus, to prevent oxidative stress of cells. NAPRT1 converts Nicotinic acid (NA; niacin) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD).
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human small intestine Tissue, 1:20-1:200
Type: Primary
Antigen: NAPRT
Clonality: Monoclonal
Clone: 5D8H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse