To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Golgi SNAP Receptor Complex member 2
- Clonality:Monoclonal
- Clone:7E7C7
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:GOSR2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:Membrin|GS27|EPM6|Bos1
- Storage buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein A Purification
- Cat. no.:10087-728
- Supplier No.:66134-1-IG
Specifications
About this item
Golgi SNAP receptor complex member 2 (GOSR2), also known as GS27 or Membrin, is a member of the solube NSF attachment protein receptor (SNARE) family of vesicle docking proteins. GOSR2 is localized in Golgi apparatus. It is involved in transport of proteins from the cis/medial-Golgi to the trans-Golgi network.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200
Type: Primary
Antigen: GOSR2/Membrin
Clonality: Monoclonal
Clone: 7E7C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human