To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily D, Polypeptide 6
- Clonality:Monoclonal
- Clone:8B11D12
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:CYP2D6
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:P450C2D|CYP2D7BP|P450-DB1|CYP2DL1|P450DB1|CYP2D7AP|Cytochrome P450 2D6|CYP2D8P2|CYP2D7P2|CYPIID6|CYP2D|CPD6
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. no.:10085-518
- Supplier No.:60297-1-IG
Specifications
About this item
CYP2D6(Cytochrome P450 2D6) is also named as CYP2DL1 and belongs to the cytochrome P450 family. It is a monooxygenase that involved in the metabolism of drugs such as antiarrhythmics, adrenoceptor antagonists, and tricyclic antidepressants. CYP2D6 is responsible for the metabolism of many drugs and environmental chemicals that it oxidizes. It has 2 isoforms produced by alternative splicing with the molecular weight of 56 kDa and 50 kDa.
Western Blot: HepG2 Cells, 1:500-1:5000
Type: Primary
Antigen: CYP2D6
Clonality: Monoclonal
Clone: 8B11D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: