Order Entry
ContactUsLinkComponent
Anti-RXRA Mouse Monoclonal Antibody [clone: 4H6C4]
Anti-RXRA Mouse Monoclonal Antibody [clone: 4H6C4]
Catalog #: 10093-736
Supplier:  Proteintech
Anti-RXRA(Monoclonal)
Anti-RXRA Mouse Monoclonal Antibody [clone: 4H6C4]
Catalog #: 10093-736
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Retinoid X Receptor, Alpha
  • Clonality:
    Monoclonal
  • Clone:
    4H6C4
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Antigen symbol:
    RXRA
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    150 µL
  • Western blot:
    Yes
  • Antigen synonyms:
    ribonucleotide reductase M1 Polypeptide|NR2B1
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Protein G Purification
  • Cat. no.:
    10093-736
  • Supplier No.:
    60198-1-IG

Specifications

About this item

Retinoid X receptor alpha (RXRA). Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RAR/RXR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. The high affinity ligand for RXRs is 9-cis retinoic acid. RXRA serves as a common heterodimeric partner for a number of nuclear receptors. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence of ligand, the RXR-RAR heterodimers associate with a multiprotein complex containing transcription corepressors that induce histone acetylation, chromatin condensation and transcriptional suppression. On ligand binding, the corepressors dissociate from the receptors and associate with the coactivators leading to transcriptional activation. The RXRA/PPARA heterodimer is required for PPARA transcriptional activity on fatty acid oxidation genes such as ACOX1 and the P450 system genes. This antibody is a rabbit polyclonal antibody raised against the 350 AA of human RXRA C-terminal. RXRA is highly expressed in liver, also expressed in lung, kidney and heart.

Western Blot: Hela Cells, 1:200-1:1000

Type: Primary
Antigen: RXRA
Clonality: Monoclonal
Clone: 4H6C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse

Customers Who Bought This Also Bought