To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B-cell lymphoma 2
- Clonality:Monoclonal
- Clone:4H8C6
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:BCL2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PPP1R50|Bcl-2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic acid/ammonium sulfate precipitation
- Supplier No.:60178-1-IG
Specifications
About this item
BCL2 belongs to the Bcl-2 family. It is a suppresses apoptosis in a variety of cell systems including factor-dependent lymphohematopoietic and neural cells. BCL2 regulates cell death by controlling the mitochondrial membrane permeability. It appears to function in a feedback loop system with caspases. BCL2 inhibits caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1).
Western Blot:Jurkat Cells, 1:500-1:5000; FC:Jurkat Cells, N/A
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 4H8C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human