Anti-PCMT1 Mouse Monoclonal Antibody [clone: 3C8G8]
Catalog #: 10091-450
Supplier: Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein-L-isoaspartate(D-aspartate) O-methyltransferase
- Clonality:Monoclonal
- Clone:3C8G8
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:PCMT1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PIMT|Protein L-isoaspartyl/D-aspartyl methyltransferase|Protein-L-isoaspartate(D-aspartate) O-methyltransferase|PCMT1|Protein-beta-aspartate methyltransferase|L-isoaspartyl protein carboxyl methyltransferase
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein G Purification
- Supplier No.:60172-1-IG
Specifications
About this item
Western Blot: Raji Cells, 1:500-1:5000
Type: Primary
Antigen: PCMT1
Clonality: Monoclonal
Clone: 3C8G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Customers Who Bought This Also Bought