To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Lectin, Galactoside-binding, Soluble, 3 binding Protein
- Clonality:Monoclonal
- Clone:2D8E11
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:LGALS3BP
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:MAC-2-BP|BTBD17B|gp90|M2BP|CyCAP|TANGO10B|90K
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic acid/ammonium sulfate precipitation
- Supplier No.:60066-1-IG
Specifications
About this item
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP is a widely expressed, secreted galectin. It has been found elevated in the serum of patients with various types of breast, lung, colorectal, ovary, and endometrial cancer, and in those infected by the human immunodeficiency virus (HIV). High expression levels of LGALS3BP are associated with a shorter survival, the occurrence of metastasis or a reduced response to chemotherapy. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1. It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Immunoblotting showed that LGALS3BP is likely contained in breast milk, serum, semen, saliva, urine, and tears.
Western Blot: Human Blood Tissue, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200
Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: 2D8E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human