To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Annexin A2
- Clonality:Monoclonal
- Clone:3H1B11
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Antigen symbol:ANXA2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:chromobindin 8|LIP2|LPC2|PAP-IV|annexin-2|protein I|placental anticoagulant protein IV|annexin II|LPC2D|CAL1H|ANX2L4|chromobindin-8|lipocortin II|calpactin I heavy chain|P36|calpactin I heavy polypeptide|calpactin-1 heavy chain|ANX2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic Acid/Ammonium Sulfate Precipitation
- Cat. no.:10081-934
- Supplier No.:60051-1-IG
Specifications
About this item
Annexin A2 (ANXA2), also named Annexin-2 or Lipocortin II, is a Ca2+ binding protein that is up-regulated in virally transformed cell lines and human tumors. This calcium-regulated membrane-binding protein, whose affinity for calcium is greatly enhanced by anionic phospholipids, may cross-link plasma membrane phospholipids with actin and the cytoskeleton and be involved with exocytosis. Heat-stress response may also involve Annexin-2.
Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:500-1:5000; FC: HeLa Cells, N/A
Type: Primary
Antigen: ANXA2
Clonality: Monoclonal
Clone: 3H1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse