To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium binding Protein A11
- Clonality:Monoclonal
- Clone:1A3F1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Antigen symbol:S100 A11
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:MLN70|HEL-S-43|S100C|Calgizzarin|Protein S100C|Protein S100-A11|S100A11|S100 calcium-binding protein A11
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic acid/ammonium sulfate precipitation
- Supplier No.:60024-1-IG
Specifications
About this item
S100A11,also named as MLN 70, Calgizzarin and S100C, Belongs to the S-100 family. It facilitates the differentiation and the cornification of keratinocytes. The naming of S100A11 systematically arises from its membership of the S100 protein family, named after their solubility in 100% saturated ammonium sulfate solution. First discovered in 1989, S100A11 has since been proposed to have direct biological functions in an assortment of physiological processes such as endo- and exocytosis, regulation of enzyme activity, cell growth and regulation, apoptosis and inflammation.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human skin, 1:20-1:200
Type: Primary
Antigen: S100 A11
Clonality: Monoclonal
Clone: 1A3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse