To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, alpha 10 (neuronal)
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CHRNA10
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:NACHRA10
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10084-738
- Supplier No.:55291-1-AP
Specifications
About this item
CHRNA10, also named as NACHRA10, belongs to the ligand-gated ion channel (TC 1.A.9) family, Acetylcholine receptor (TC 1.A.9.1) subfamily and Alpha-10/CHRNA10 sub-subfamily. It is an ionotropic receptor with a probable role in the modulation of auditory stimuli. In the ear, CHRNA10 may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. It may protect against acoustic trauma. This antibody is specific to CHRNA10.
Western Blot: Raji Cells, 1:200-1:1000
Type: Primary
Antigen: CHRNA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: