To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:LNPEP
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10089-744
- Supplier No.:55154-1-AP
Specifications
About this item
LNPEP, also named as OTASE, IRAP, P-LAP and Otase, belongs to the peptidase M1 family. It may be involved in the inactivation of neuronal peptides in the brain. LNPEP cleaves Met-enkephalin and dynorphin. It binds angiotensin IV and may be the angiotensin IV receptor in the brain. It can be detected a protein with molecular mass of 165 kDa in several human tissues including heart, small intestine, kidney and 140 kDa in brain. LNPEP contains 18 potential N-glycosylation sites so that the differential usage of the glycosylation site in the brain causes the difference in the molecular mass of the enzyme.. Two forms of LNPEP are purified from human placenta: one is a monomeric, 61-kDa protein and the other is a heterodimeric protein with 64- and 97-kDa subunits. It can form a homodimer of 280 kDa. This antibody is special to LNPEP.
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200
Type: Primary
Antigen: LNPEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human