To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Discs, Large (Drosophila) Homolog-Associated protein 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:DLGAP3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DAP-3|SAPAP3|DAP3|Disks large-associated protein 3
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10085-898
- Supplier No.:55056-1-AP
Specifications
About this item
DLGAP3, also named as DAP3, belongs to the SAPAP family. It may play a role in the molecular organization of synapses and neuronal cell signaling. DLGAP3 could be an adapter protein linking ion channel to the subsynaptic cytoskeleton. It may induce enrichment of PSD-95/SAP90 at the plasma membrane. The antibody has no cross reaction to other DLGAP members.
Western Blot: Human Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: DLGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse