To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protein 161A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TMEM161A
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:T161A_HUMAN|Transmembrane protein 161A.|tmem161a|AROS-29
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Supplier No.:24898-1-AP
Specifications
About this item
TMEM161A also named as transmembrane protein 161A is a 479 amino-acid protein, which belongs to TMEM161 family. The molecular weight of TMEM161A is 56 kDa. TMEM161A is multi-pass membrane protein and localizes on the cell membrane. It may involve in the negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage and cellular response to oxidative stress.
Western Blot: MCF-7 Cells, 1:500-1:5000
Type: Primary
Antigen: TMEM161A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human