To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bromodomain Containing 9
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BRD9
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:bromodomain-containing protein 9|sarcoma antigen NY-SAR-29|FLJ43530|LAVS3040|PRO9856|DKFZp434D0711|rhabdomyosarcoma antigen MU-RMS-40.8|DKFZp686L0539|UNQ3040/PRO9856
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10083-676
- Supplier No.:24785-1-AP
Specifications
About this item
BRD9, also named as MU-RMS-40.8, may play a role in chromatin remodeling and regulation of transcription. BRD9 has six isoforms with MW 67 kDa, 23 kDa, 53 kDa, 26 kDa, 56 kDa and 61 kDa.
Western Blot: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: BRD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human