To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) Polypeptide G(32kDa)
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:POLR3G
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:RPC32|RPC7
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10092-580
- Supplier No.:24701-1-AP
Specifications
About this item
POLR3G also named as RNA polymerase III subunit C7 is a 223 amino acid protein, which belongs to eukaryotic RPC7 RNA polymerase subunit family. The molecular weight of POLR3G is 26 kDa. POLR3G localizes in the nucleus and is a component of the RNA polymerase III complex, which may be involved either in the recruitment and stablization of the subcomplex within RNA polymerase III, or in stimulating catalytic functions of other subunits during initiation.
Western Blot: HeLa Cells, 1:500-1:5000; IP: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: POLR3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human