Anti-GPR58 Rabbit Polyclonal Antibody
Catalog #: 10087-796
Supplier: Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:G Protein-coupled Receptor 58
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GPR58
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:GPR 58|phBL5|G protein coupled receptor 58|TAAR2|Trace amine associated receptor 2.|GPCR 58
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10087-796
- Supplier No.:24609-1-AP
Specifications
About this item
Western Blot:rat Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: GPR58/TAAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought