To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 67
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:TRIM67
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-360
- Supplier No.:24369-1-AP
Specifications
About this item
TRIM67(Tripartite motif-containing protein 67) is also named as TNL and belongs to the belongs to the TRIM/RBCC family. The deduced protein contains an N-terminal RING finger domain, 2 B-boxes, a coiled-coil region, a COS box, an FN3 domain, and a B30.2 domain. TRIM67 shares 65% amino acid identity with TRIM9. This protein has 2 isoforms produced by alternative splicing.
Western Blot: HepG2 Cells, 1:200-1:2000
Type: Primary
Antigen: TRIM67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse