To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Catalase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CTA1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Supplier No.:21260-1-AP
Specifications
About this item
CAT(Catalase) belongs to the catalase family. It is involved in catabolic pathway of activated oxygen species and free radical detoxification and the catalase overexpression has a protective role against UVB irradiation by preventing DNA damage mediated by the late ROS increase. Mammalian catalase of approximately 240 kD occurs as a complex of 4 identical subunits, each of which contains 526 amino acid residues. Defects in CAT are the cause of acatalasemia (ACATLAS). This antibody is specific to CAT.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: CTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: