To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Beta
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Monkey,Human,Rat,Mouse
- Antigen symbol:ACTB
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10081-224
- Supplier No.:20536-1-AP
Specifications
About this item
Beta actin, also named as ACTB and F-Actin, belongs to the actin family. Actins are highly conserved globular proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells. At least six isoforms of actins are known in mammals and other vertebrates: alpha (ACTC1, cardiac muscle 1), alpha 1 (ACTA1, skeletal muscle) and 2 (ACTA2, aortic smooth muscle), beta (ACTB), gamma 1 (ACTG1) and 2 (ACTG2, enteric smooth muscle). Beta and gamma 1 are two non-muscle actin proteins. Most actins consist of 376aa, while ACTG2 (rich in muscles) has 375aa and ACTG1(found in non-muscle cells) has only 374aa. Beta actin has been widely used as the internal control in RT-PCR and Western Blotting as a 42-kDa protein. This antibody was generated against N-terminal region of human beta actin protein and can cross-react with other actins.
- IF: HepG2 Cells, 1:10-1:100; Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Colon Tissue, 1:20-1:200
Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish, Monkey