To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Small Nuclear Ribonucleoprotein Polypeptide E
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SNRPE
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-954
- Supplier No.:20407-1-AP
Specifications
About this item
The snRNP E (SNRPE)protein is one of several proteins associated with the U family of small nuclear RNAs that are involved in RNA processing. It functions in the U7 snRNP complex that is involved in histone 3'-end processing and also associated with snRNP U1, U2, U4/U6 and U5. This antibody raises against the full length of human SNRPE, and can recognize two bands of SNRPE, include a 9 kDa band(isoform) and a 11kDa band(SNRPE)
Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200
Type: Primary
Antigen: SNRPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat