To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAH (Asp-Glu-Ala-His) box polypeptide 32
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DHX32
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DHLP1|DDX32
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Supplier No.:19808-1-AP
Specifications
About this item
DHX32, a new member of the DEAH subfamily of DEAD box proteins, is a novel putative RNA helicases with an activation-dependent paten of expression, and characterized by a conserved helicase domain consisting of 7 to 8 conserved motifs. RNA helicases of the DEAD box family is are highly conserved enzymes which are required for RNA metabolism from transcription to degradation and are therefore important players in gene expression. DHX32 may play an important role in the development of colorectal cancer . This is a rabbit polyclonal anti-DHX32 antibody raised against part of C-terminal chain of human DHX32.
Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200
Type: Primary
Antigen: DHX32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat