To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tubulointerstitial Nephritis Antigen
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:TINAG
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Supplier No.:19747-1-AP
Specifications
About this item
TINAG belongs to the peptidase C1 family. TINAG mediates adhesion of proximal tubule epithelial cells via integrins alpha3-beta1 and alphaV-beta3. It is a non catalytic peptidase C1 family protein. Antibodies against TINAG are found in sera of patients with tubulointerstitial nephritis, a rare autoimmune disorder that causes acute and chronic renal injury.It has 2 isoforms produced by alternative splicing. The antibody is specific to TINAG.
Western Blot: Mouse small intestine Tissue, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200
Type: Primary
Antigen: TINAG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse