To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Prenylcysteine oxidase 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PCYOX1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10091-960
- Supplier No.:18868-1-AP
Specifications
About this item
PCYOX1(Prenylcysteine oxidase 1) is also named as KIAA0908, PCL1, prenylcysteine lyase and belongs to the prenylcysteine oxidase family. The deduced 505-amino acid protein contains a predicted signal sequence, several transmembrane regions, and four potential N-glycosylation sites. It is a glycosylated enzyme involved in prenylated protein catabolism, releasing a free cysteine and a hydrophobic isoprenoid product. Immunofluorescence studies of PCYOX1 showed a nonnuclear punctate pattern, and PCYOX1 colocalized with lysosomal marker LAMP1 in HEK293 cells. The enzyme is isolated from bovine brain membranes and exhibits an apparent molecular mass of 63 kDa.
IHC: Human Colon Tissue, 1:20-1:200
Type: Primary
Antigen: PCYOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat