To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Rho GTPase activating Protein 24
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Antigen symbol:ARHGAP24
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10083-076
- Supplier No.:18834-1-AP
Specifications
About this item
ARHGAP24, also named as FILGAP, is involved in cell polarity, cell morphology and cytoskeletal organization. ARHGAP24 acts as a GTPase activator for the Rac-type GTPase by converting it to an inactive GDP-bound state. ARHGAP24 controls actin remodeling by inactivating Rac downstream of Rho leading to suppress leading edge protrusion and promotes cell retraction to achieve cellular polarity. ARHGAP24 can suppress the activity of RAC1 and CDC42 in vitro. Its overexpression induces cell rounding with partial or complete disruption of actin stress fibers and formation of membrane ruffles, lamellipodia, and filopodia. Isoform 2 is a vascular cell-specific GAP involved in modulation of angiogenesis. The antibody may recognize all the isoforms of ARHGAP24 except isoform 5.
Western Blot: Mouse ovary Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200
Type: Primary
Antigen: ARHGAP24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Zebrafish