To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Phosphatase 1, Regulatory Subunit 9A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PPP1R9A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:Neurabin 1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Supplier No.:18801-1-AP
Specifications
About this item
PPP1R9A, also named as KIAA1222 and NEB1, binds to actin filaments (F-actin) and shows cross-linking activity. PPP1R9A has been found to be concentrated in the synapse of developed neurons and in the lamellipodia of the growth cone during neuron development.PPP1R9A may be involved in neurite formation. PPP1R9A inhibits protein phosphatase 1-alpha activity.
Western Blot:PC-3 Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100
Type: Primary
Antigen: PPP1R9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat