To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Proteasome (prosome macropain) Activator Subunit 4
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PSME4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PA200
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Supplier No.:18799-1-AP
Specifications
About this item
PSME4, like PSME1, 2, and 3, encodes proteasome activator complex subunit 4 also named proteasome activator 200 kDa (PA200). PA200 activates proteasomal cleavage of peptides in an energy-independent manner. After exposure of cells to ionizing radiation, PA200 forms nuclear foci and enhances proteasome activity independent of the stage of cell cycle arrest. PA200 also may be involved in spermatogenesis. The antibody is expected to recognize all the 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140-kDa.
Western Blot: HEK-293 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IP: HEK-293 Cells, 1:200-1:2000
Type: Primary
Antigen: PSME4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat