To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DHX9
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:17721-1-AP
Specifications
About this item
RNA helicases play important roles in transcription, RNA processing, translation, and RNA replication. DEAD box proteins are putative RNA helicases that have a characteristic Asp-Glu-Ala-Asp (DEAD) box as 1 of 8 highly conserved sequence motifs. DHX9 a member of the DEAH family of proteins, which possess a double-stranded RNA-binding domain (dsRBD) and a helicase domain . It unwinds double-stranded DNA and RNA in a 3' to 5' direction. Alteration of secondary structure of DHX9 may subsequently influence interactions with proteins or other nucleic acids. It is also a component of the CRD-mediated complex that promotes MYC mRNA stability. In addition, it is involved with LARP6 in the stabilization of type I collagen mRNAs for CO1A1 and CO1A2.
Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200
Type: Primary
Antigen: DHX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat