To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bobby Sox Homolog
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BBX
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:MDS001|HBP2|HSPC339|ARTC1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:17254-1-AP
Specifications
About this item
BBX, also named HBP2, is a protein with a high mobility group (HMG) domain that belongs to the Sox (Sry-related HMG box) family of transcription factors. As a DNA-binding domain in HBP2, HMG is essential for its function in Schizosaccharomyces pombe. HBP2 activates CdcIO-dependent transcription at G1/s and allows growth of cdc10-129, resldelta and rep2delta mutant cells at the restrictive temperature
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human ovary, 1:20-1:200
Type: Primary
Antigen: BBX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat