To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Filament Associated Protein 1-like 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:AFAP1L2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:afap1l2.|AF1L2_HUMAN|KIAA1914|AFAP1-like protein 2|Actin filament-associated protein 1-like 2|XB130
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:17183-1-AP
Specifications
About this item
AFAP1L2, also named as Actin filament-associated protein 1-like 2 or XB130, is a 818 amino acid protein, which contains two PH domains. AFAP1L2 is detected in spleen and thyroid, and at lower levels in kidney, brain, lung and pancreas. AFAP1L2 localizes in the cytoplasm and may play a role in a signaling cascade by enhancing the kinase activity of SRC. AFAP1L2 promotes thyroid tumor growth, and its expression in these cancer cells may affect cell proliferation and survival by controlling the expression of multiple genes, especially transcription regulators.
Western Blot: Mouse Thymus Tissue, 1:500-1:5000
Type: Primary
Antigen: AFAP1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat