To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Sperm Associated Antigen 16
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SPAG16
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:WDR29|PF20|Pf20 protein homolog|SPAG16|Sperm-associated antigen 16 protein
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:16883-1-AP
Specifications
About this item
SPAG16 (also known as PF20) is an orthologue of Chlamydomonas PF20, a component of the alga axoneme central apparatus that is required for flagellar motility. SPAG16 is highly expressed in the testis, low abundant or absent from other tissues. The SPAG16 gene encodes two major proteins, a 71-kDa SPAG16L and a smaller 35-kDa SPAG16S nuclear protein that represents the C-terminus of SPAG16L. SPAG16L was detected abundantly in the cytoplasm, while SPAG16S was detected in both cytoplasm and nucleus. This antibody was raised against the N-terminal region of SPAG16 and mainly recognizes SPAG16L.
Western Blot:rat Testis Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200
Type: Primary
Antigen: SPAG16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat