To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Na+/K+ transporting, alpha 2 Polypeptide
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ATP1A2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:ATP1A2
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:16836-1-AP
Specifications
About this item
ATP1A2 (Na+/K+-ATPase α-2 subunit) is the catalytic component of the active enzyme Na+/K+-ATPase, which catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. The Na+/K+-ATPase is composed of a larger catalytic α-subunit (~110 kDa) and a small β-subunit (~55 kDa). The α subunit has four isoforms identified to date: α1, α2, α3 and α4.The α1 isoform is expressed ubiquitously but the α2 isoform is present largely in the skeletal muscle, heart and vascular smooth muscle. The α3 isoform is found almost exclusively in neurons and ovaries. The α4 isoform is expressed in sperm. This antibody was raised against the internal region of the human ATP1A2 and can recognizes all the isoforms of α subunit. The 65kDa band detected occasionally may be the degradation product of ATP1A2.
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; FC: HeLa Cells, N/A
Type: Primary
Antigen: ATP1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat