To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Claudin 4
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CLDN4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10084-844
- Supplier No.:16195-1-AP
Specifications
About this item
Claudins are a family of proteins that are the most important components of the tight junctions, where they establish the paracellular barrier that controls the flow of molecules in the intercellular space between the cells of an epithelium. 23 claudins have been identified. They are small (20-27 kDa) proteins with very similar structure. They have four transmembrane domains, with the N-terminus and the C-terminus in the cytoplasm. This antibody is specifically against claudin 4.
Western Blot: Mouse small intestine Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200
Type: Primary
Antigen: CLDN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: