To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glycyl-tRNA synthetase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GARS
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:GARS|HMN5|DSMAV|SMAD1|Glycine--tRNA ligase|CMT2D|GlyRS
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10087-392
- Supplier No.:15831-1-AP
Specifications
About this item
GARS(glycinamide ribonucleotide synthetase) perform an essential function in protein synthesis by catalyzing the esterification of an amino acid to its cognate tRNA.GARS, however, is a ubiquitously expressed α2 homodimer responsible for charging tRNA molecules with glycine, this enzyme is essential for protein synthesis in all cells and tissues.It can exist as a oligomer with 250KDa.
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human skin cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: GARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat