To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Dehydrogenase/reductase (SDR family) Member 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:DHRS2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:SDR25C1|HEP27
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-822
- Supplier No.:15735-1-AP
Specifications
About this item
DHRS2(Dehydrogenase/reductase SDR family member 2) is also named as dicarbonyl reductase HEP27, Protein D and belongs to the short-chain dehydrogenases/reductases (SDR) family. It displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates and there is no reductase activity displayed in vitro with steroids, retinoids and sugars as substrates. This protein also may inhibit cell replication. It has 2 isoforms produced by alternative splicing with the molecular weight of 27 kDa and 29 kDa.
Western Blot: Human Liver Tissue, 1:500-1:5000
Type: Primary
Antigen: DHRS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human