To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase)
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NDUFS7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:NADH-coenzyme Q reductase|FLJ45860|20kDa (NADH-coenzyme Q reductase)|mitochondrial respiratory chain|FLJ46880|MGC120002|NADH dehydrogenase (ubiquinone) Fe-S protein 7|complex I-20kD|MY017|mitochondrial|NDUFS7|PSST|CI-20KD|complex I|20-KD subunit|NADH:ubiquinone oxidoreductase PSST subunit|NADH-ubiquinone oxidoreductase Fe-S protein 7|NADH dehydrogenase [ubiquinone] iron-sulfur protein 7
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10090-988
- Supplier No.:15728-1-AP
Specifications
About this item
NDUFS7(NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial) protein is one of the most conserved subunits of mitochondrial respiratory chain complex I and plays a central role in the interaction with the electron acceptor ubiquinone and in the proton-translocating mechanism.Defects in NDUFS7 are a cause of Leigh syndrome (LS) and mitochondrial complex I deficiency (MT-C1D).
Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human Liver Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100
Type: Primary
Antigen: NDUFS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat