To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bile salt-activated lipase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BSDL
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:15384-1-AP
Specifications
About this item
Bile salt-activated lipase, previously named cholesterol esterase or bile salt-stimulated (or dependent) lipase, is a 74 kDa lipolytic enzyme capable of hydrolyzing cholesteryl esters, tri-, di-, and mono-acylglycerols, phospholipids, lysophospholipids, and ceramide. The same enzyme is present as a major protein in milk and as a minor constituent in liver, activated macrophages, and endothelial cells. Defects in CEL are a cause of maturity-onset diabetes of the young type 8 with exocrine dysfunction (MODY8). The full length protein has 11 glycosylation sites. It has 2 isoforms produced by alternative splicing.
Western Blot: Human Liver Tissue, 1:500-1:5000
Type: Primary
Antigen: BSDL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human