To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Galactosamine-6 Sulfatase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GALNS
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:GALNAC6S|GAS|GalN6S|MPS4A
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:14933-1-AP
Specifications
About this item
GALNS(N-acetylgalactosamine-6-sulfatase) is also named as chondroitinase and belongs to the sulfatase family. It is one of sulfatases required to degrade glycosaminoglycans (GAGs), keratan sulfate (KS) and chondroitin-6-sulfate (C6S) and the enzyme is a dimer derived from two 60 kDa polypeptides, each of which is processed to 40 kDa and 15 kDa polypeptide subunits linked by disulfide bonds. The deduced 522-residue protein is composed of a 26-amino acid N-terminal signal peptide and a mature polypeptide of 496 amino acid residues, including 2 potential asparagine-linked glycosylation sites. Defects in GALNS are the cause of mucopolysaccharidosis type 4A (MPS4A), also known as Morquio A syndrome.
Western Blot: Human Liver Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200
Type: Primary
Antigen: GALNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat