To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Quinoid Dihydropteridine Reductase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:QDPR
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PKU2|DHPR|SDR33C1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:14908-1-AP
Specifications
About this item
Dihydropteridine reductase (QDPR), also named as DHPR and HDHPR, is an essential enzyme in the hydroxylating system of the aromatic amino acids, since it catalyses the regeneration of tetrahydrobiopterin (BH4), the natural cofactor of phenylalanine, tyrosine, and tryptophan hydroxylases, from the quininoid-dihydrobiopterin produced in these coupled reactions. The QDPR protein is active as a dimer, with a subunit Mr of 26 kDa. This protein belongs to the short-chain dehydrogenases/reductases (SDR) family. Defects in QDPR are the cause of BH4-deficient hyperphenylalaninemia type C (HPABH4C).
Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200
Type: Primary
Antigen: QDPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat