To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcium/Calmodulin-Dependent Protein Kinase Kinase 1, alpha
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CAMKK1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:MGC34095|Ca2+/calmodulin-dependent protein kinase kinase alpha|CaM-kinase kinase 1|CaMKK 1|Calcium/calmodulin-dependent protein kinase kinase alpha|Calcium/calmodulin dependent protein kinase kinase alpha|CaMKK|CaMKK alpha|Calcium/calmodulin dependent protein kinase kinase 1 alpha|DKFZp761M0423|CaM-kinase IV kinase|Camkk1|Calcium/calmodulin dependent protein kinase kinase|CAMKK alpha protein|KKCC1_HUMAN|CaM-KK alpha|CAMKKA|Calcium/calmodulin-dependent protein kinase kinase 1|CaM-kinase kinase alpha|CaM-KK 1|CaM kinase kinase alpha|OTTMUSP00000006419.|Calcium/calmodulin dependent protein kinase kinase 1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:13624-1-AP
Specifications
About this item
Calcium/calmodulin-dependent protein kinase kinase 1 catalyzes the phosphorylation and activation of Ca(2+)/calmodulin kinase (CaMK) as an upstream component of the CaMK cascade that has been implicated in neuronal gene transcription, synaptic plasticity, and long-term memory consolidation .CAMKK1 has two isoforms of 56kda and 58kda and involved in regulating cell apoptosis, promotes cell survival by phosphorylating AKT1/PKB that inhibits pro-apoptotic BAD/Bcl2-antagonist of cell death .Researches showed that Camkk1 plays a selective role in contextual fear memory.This 66-68kda protein can be phosphorylated(probably antophosphorylation).This antibody recognize the N-terminal of CAMKK1 .
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200
Type: Primary
Antigen: CAMKK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat