To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adenosine Deaminase, tRNA-specific 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ADAT2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:ADAT2
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:13621-1-AP
Specifications
About this item
ADAT2(tRNA-specific adenosine deaminase 2) is also named as DEADC1 and Belongs to the cytidine and deoxycytidylate deaminase family. It is a 24-kDa molecular mass protein that harbors all the conserved motifs required for deamination. ADAT2 and ADAT3 can function as a heterodimer which catalyses inosine formation at the wobble position (position 34) in eukaryotic tRNAs. It has 2 isoforms produced by alternative splicing.
Western Blot:A431 Cells, 1:200-1:2000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200
Type: Primary
Antigen: ADAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat