To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 6
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SIRT6
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:SIR2-like protein 6|Mono-ADP-ribosyltransferase sirtuin-6|Silent information regulator 2-like protein 6.
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:13572-1-AP
Specifications
About this item
SIRT6, also named as SIR2L6, belongs to the sirtuin family. SIRT6 is a member of a highly conserved family of NAD+-dependent deacetylases with various roles in metabolism, stress resistance, and life span. It has deacetylase activity towards 'Lys-9' and 'Lys-56' of histone H3. SIRT6 modulates acetylation of histone H3 in telomeric chromatin during the S-phase of the cell cycle. It deacetylates 'Lys-9' of histone H3 at NF-kappa-B target promoters and may down-regulate the expression of a subset of NF-kappa-B target genes. SIRT6 may be required for the association of WRN with telomeres during S-phase and for normal telomere maintenance. It is required for genomic stability. And it is required for normal IGF1 serum levels and normal glucose homeostasis. It modulates cellular senescence and apoptosis. It regulates the production of TNF protein. SIRT6 acts as a histone deacetylase to directly repress multiple glycolytic genes. SIRT6 also functions as a corepressor of the transcription factor Hif1α. Loss of SIRT6 increases glycolysis and diminishes mitochondrial respiration. SIRT6 plays a key role in regulating glucose homeostasis.
- Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Brain, 1:20-1:200
Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat