To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ubiquilin 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UBQLN3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10081-318
- Supplier No.:13568-1-AP
Specifications
About this item
UBQLN3(Ubiquilin-3) may interact with the testis-specific cyclin A1 and regulate cell cycle progression during spermatogenesis, as does frog Xdrp1, with which UBQLN3 shares sequence similarity.UBQLN3 expression is not observed in mouse eye, thyroid, pancreas,smooth muscle, ovary, submaxillary gland, uterus, and epididymis by northern blot.
Western Blot: Human Testis Tissue, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:2000
Type: Primary
Antigen: UBQLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat