To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CNAP1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:13382-1-AP
Specifications
About this item
NCAPD2 is an essential component of the highly conserved condensin complex required for mitotic chromosome condensation and for the correct attachment between chromosome kinetochores and microtubules of the mitotic spindle . NCAPD2,also known as CEP250 or C-Nap1, is a centrosomal protein and the presumed substrate of Nek2 and PP1. The function of C-Nap1 in centriolar cohesion may be regulated by its own phosphorylation status, which depends on the balance between Nek2 and PP1 activities. Studies by immunoelectron microscopy found that C-Nap1 located only at the proximal ends of, but not between, the two centrioles .
Western Blot: HEK-293 Cells, 1:200-1:2000; IF: Hela Cells, 1:20-1:200
Type: Primary
Antigen: CNAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: