To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Corneodesmosin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:CDSN
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Antigen synonyms:HYPT2|HTSS|S|PSS1|PSS|HTSS1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-550
- Supplier No.:13184-1-AP
Specifications
About this item
CDSN, also named as S protein, is important for the epidermal barrier integrity. CDSN is a unique extracellular 52- to 56-kDa glycoprotein found in corneodesmosomes. Pathogenic mutations in CDSN have been linked in humans to genodermatoses affecting both the hair follicle and the epidermis. Inflammatory peeling skin syndrome is caused a novel mutation in CDSN.
IHC: Human skin cancer, 1:20-1:200
Type: Primary
Antigen: CDSN
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: