To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAH (Asp-Glu-Ala-His) box polypeptide 36
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DHX36
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DHX36|Probable ATP-dependent RNA helicase DHX36|RHAU|DDX36|KIAA1488|DEAH box protein 36|MLEL1|G4R1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-840
- Supplier No.:13159-1-AP
Specifications
About this item
DHX36, also known as RNA associated with AU-rich element (RHAU) and G4R1, is an adenosine triphosphate (ATP)-dependent RNA helicase that belongs to the DExH/D family of RNA modifying enzymes. Proteins in this family involve in a wide range of cellular functions including RNA splicing, ribosome assembly, messenger RNA (mRNA) stability, microRNA processing, ribonucleoprotein remodeling and RNA trafficking. DHX36 is an RNA helicase that has specificity for DNA and RNA G4-quadruplexes, and resolves G4 structures into separate nucleotide strands for metabolic processing of G-rich sequence.
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human prostate Tissue, 1:20-1:200
Type: Primary
Antigen: DHX36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat