To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SECISBP2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-274
- Supplier No.:12798-1-AP
Specifications
About this item
Selenium (Se) is an essential trace element required for the biosynthesis of selenoproteins, and selenocysteine insertion sequence (SECIS) binding protein 2 (SECISBP2, or SBP2) represents a key trans-acting factor for the cotranslational insertion of selenocysteine into selenoproteins. Defects in SBP2 are a cause of abnormal thyroid hormone metabolism (ATHYHM) associated with a reduction in type II iodothyronine deiodinase activity. Mutations in this gene have been associated with a reduction in activity of a specific thyroxine deiodinase, a selenocysteine-containing enzyme, and abnormal thyroid hormone metabolism. Cells depleted of SBP2 have increased levels of ROS, which lead to cellular oxidative stress manifested as DNA lesions, stress granules, and lipid peroxidation, induction of caspase- and cytochrome c-dependent apoptosis, indicating that SBP2 is required for protection against ROS-induced cellular damage and cell survival.
Western Blot: HeLa Cells, 1:500-1:5000; IP: Mouse Testis Tissue, 1:200-1:2000
Type: Primary
Antigen: SECISBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat