To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Coagulation Factor 12
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:F12
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:HAF|Coagulation Factor XII|HAE3|Hageman Factor|HAEX
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:12551-1-AP
Specifications
About this item
Factor XII (F XII, Hageman factor) is a 80kD, single chain glycoprotein that circulates in blood as an inactive zymogen. F XII plays an important role in blood coagulation, fibrinolysis, and kinin generation. When plasma comes into contact with negatively charged surfaces such as glass, kaolin, or dextran sulfate, F XII is bound to the surface and undergoes limited proteolysis by plasma kallikrein, resulting in two active enzyme forms, ɑ-F XIIa and β-F XIIa. ɑ-F XIIa consisting of a 28kD light chain containing the protease domain, and a 52kD heavy chain containing the anionic surface-binding domain. β-F XIIa consists of two disulfide-linked chains, a 2 kD light chain and a 28 kD heavy chain containing the catalytic domain. Defects in Factor XII do not cause any clinical symptoms and the sole effect is that whole-blood clotting time is prolonged.
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200
Type: Primary
Antigen: F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat