To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAH (Asp-Glu-Ala-His) box helicase 15
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DHX15
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PRP43|PRPF43|DBP1|PrPp43p|HRH2|DDX15
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-830
- Supplier No.:12265-1-AP
Specifications
About this item
DHX15, also named as DBP1 and DDX15, is a DExD/H-box protein that has been identified in spliceosome, through its carboxyl (C)-terminal G-patch domain and stimulates the helicase activity of DHX15. DHX15 is a Pre-mRNA processing factor involved in disassembly of spliceosomes after the release of mature mRNA. The MW of DHX15 is about 90-95kd.
Western Blot:NIH/3T3 Cells, 1:200-1:2000
Type: Primary
Antigen: DHX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat